missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPSB3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531502
This item is not returnable.
View return policy
Description
Recommended positive controls: U87-MG, mouse brain. Predicted reactivity: Mouse (97%), Rat (97%), Xenopus laevis (84%), Rhesus Monkey (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May be a substrate recognition component of a SCF-like ECS (Elongin BC-CUL2/5-SOCS-box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins.
Specifications
| SPSB3 | |
| Polyclonal | |
| Unconjugated | |
| SPSB3 | |
| 2310012N15Rik; 3300001M01Rik; C16orf31; Kiaa4204; mKIAA4204; splA/ryanodine receptor domain and SOCS box containing 3; SPRY domain-containing SOCS box protein 3; SPRY domain-containing SOCS box protein SSB-3; SPSB3; SSB3; SSB-3; Tce1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79043, 90864 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q571F5, Q6PJ21 | |
| SPSB3 | |
| Recombinant protein encompassing a sequence within the center region of human SPSB3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction