missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SPRYD4 Polyclonal Antibody

Product Code. 12809322
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12809322 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 12809322 missing translation for 'mfr' Invitrogen™ missing translation for 'supplierNo' PA521174

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is mouse kidney tissue lysate. PA5-21174 can be used with blocking peptide PEP-1288.

The SPRY domain-containing protein 4 (SPRYD4) is a member of a family of proteins whose sole common characteristic is the presence of a SPRY domain. SPRY domains are structural domains that were first described in the fungal Dictyostelium discoideum tyrosine kinase spore lysis A. In most systems SPRY domains provide binding sites for regulatory proteins or intramolecular binding sites that maintain the structural integrity of a protein. SPRYD4 is ubiquitously expressed and is most abundant in kidney, brain, bladder, thymus and stomach. Little is known of the function of the SPRYD4 protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPRYD4
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SPRYD4
Gene Accession No. Q4FZT8, Q8WW59, Q91WK1
Gene Alias 4633402N23Rik; AA114801; RGD1306649; SPRY domain containing 4; SPRY domain-containing protein 4; Spryd4
Gene Symbols SPRYD4
Host Species Rabbit
Immunogen An 18 amino acid peptide near the center of human SPRYD4.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283377, 288772, 66701
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.