missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SPRYD2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12859312
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12859312 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12859312 Supplier Invitrogen™ Supplier No. PA521170

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is mouse heart tissue lysate. PA5-21170 can be used with blocking peptide PEP-1284.

SPRYD2, also known as Myospryn, was originally identified as the muscle-specific partner of dysbindin and as a Mef-2 target gene. It is a large scaffolding protein localized to the Z-disc/costamere region of striated muscle. SPRYD2 includes a noncanonical tripartite motif (TRIM-like) that lacks the RING domain but consists of a B-box coiled coil (BBC), fibronectin 3 (FN3) repeats, and SPRY domains. SPRYD2 interacts with desmin and calcineurin, and it has been suggested to play a role in the biogenesis of lysosome and negatively regulates slow-fiber-type transformation and skeletal muscle regeneration. SPRYD2 is dysregulated in Duchenne muscular dystrophy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPRYD2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CMYA5
Gene Accession No. Q70KF4, Q8N3K9
Gene Alias 2310076E16Rik; 2310076E21Rik; AA420382; AI504003; C5orf10; cardiomyopathy associated 5; cardiomyopathy-associated protein 5; CMYA5; DTNBP2; Dystrobrevin-binding protein 2; genethonin-3; heart and skeletal muscle-specific and sprouty domain-containing; myospryn; SPRY domain-containing protein 2; SPRYD2; sr553; Srfsd; stretch response protein 553; stretch-response protein 553; stretch-responsive fibronectin protein type 3; Tims; TRIM76; TRIM-like protein; tripartite motif-containing 76; tripartite motif-containing protein 76
Gene Symbols CMYA5
Host Species Rabbit
Immunogen An 18 amino acid peptide near the carboxy terminus of human SPRYD2.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 202333, 688915, 76469
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.