missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPRY4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2032
This item is not returnable.
View return policy
Description
SPRY4 Polyclonal specifically detects SPRY4 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SPRY4 | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| protein sprouty homolog 4, sprouty homolog 4 (Drosophila), spry-4 | |
| Rabbit | |
| 32.54 kDa | |
| 0.1 mg | |
| Growth and Development, Neuronal Cell Markers, Signal Transduction | |
| 81848 | |
| Human, Mouse | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.1 mg/ml | |
| Western Blot 1:100-1:1000, ELISA 1:3000-1:12000, Immunohistochemistry 1 ug/ml-10 ug/ml, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 2.5mg/ml | |
| Q9C004 | |
| SPRY4 | |
| This affinity purified SPRY4 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a C-Terminal region near amino acids 275-299 of Human SPRY4 protein. (Uniprot: Q9C004) | |
| Affinity Purified | |
| RUO | |
| Primary | |
| A BLAST analysis was used to suggest reactivity with this protein from human and chimpanzee based on 100% homology for the immunogen sequence. | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction