missing translation for 'onlineSavingsMsg'
Learn More

SPRY3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18333786 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18333786 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18333786 Supplier Novus Biologicals Supplier No. NBP309277100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SPRY3 Polyclonal specifically detects SPRY3 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPRY3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS buffer, 2% sucrose
Gene Alias Antagonist Of FGF Signaling, HSPRY3, Protein Sprouty Homolog 3, sprouty (Drosophila) homolog 2, Sprouty Homolog 3 (Drosophila), spry-3
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the N terminal region of human SPRY3 (NP_005831). Peptide sequence QLQPLPQHLSQSSIASSMSHSTTASDQRLLASITPSPSGQSIIRTQPGAG
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10251
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.