missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPRY3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17932-1-AP-150UL
This item is not returnable.
View return policy
Description
May function as an antagonist of fibroblast growth factor (FGF) pathways and may negatively modulate respiratory organogenesis.Specifications
| SPRY3 | |
| Polyclonal | |
| Unconjugated | |
| SPRY3 | |
| HSPRY3, Protein sprouty homolog 3, sprouty homolog 3 (Drosophila), Spry 3, SPRY3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10251, 236576, 498159 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43610 | |
| Spry3 | |
| SPRY3 Fusion Protein Ag12364 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction