Learn More
SPRY1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA543582
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: PTAILSLDQI KAIRGSNEYT EGPSVVKRPA PRTAPRQEKH ERTHEIIPIN Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%
SPRY1 Polyclonal antibody specifically detects SPRY1 in Human, Mouse samples. It is validated for Western Blot.Specifications
SPRY1 | |
Polyclonal | |
Unconjugated | |
SPRY1 | |
hSPRY1; Protein sprouty homolog 1; sprouty 1; sprouty homolog 1 (Drosophila); sprouty homolog 1, antagonist of FGF signaling; sprouty homolog 1, antagonist of FGF signaling (Drosophila); sprouty RTK signaling antagonist 1; sprouty, Drosophila, homolog of, 1 (antagonist of FGF signaling); sprouty1; SPRY1; spry-1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10252, 24063 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O43609, Q9QXV9 | |
SPRY1 | |
Synthetic peptide directed towards the N-terminal of human SPRY1 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |