Learn More
Replacement O-Rings for Fisherbrand™ Mini Centrifuge
Product Code. 15435380 Shop All Thermo Scientific CO₂ Incubators Products
Click to view available options
For Use With (Equipment):
Fisherbrand Mini Centrifuge (16617645)
Unit Size:
Each
Product Code. 15435380
Brand: Heathrow Scientific HS120148
Only null left
Add to basket
Add to basket
Act as tiny hinges on the Mini-Centrifuge lid. Replacement O-Rings also help to keep the lid in place and prevents squeaking. For use with Fisherbrand Mini Centrifuge (16617645).
  • For use with Fisherbrand™ Mini-Centrifuge
  • Latex Free
  • Available in packs of 10
Specifications
For Use With (Equipment) Fisherbrand Mini Centrifuge (16617645)
Quantity 10 Pack
Product Type O-Ring
Material Latex-free rubber