missing translation for 'onlineSavingsMsg'
Learn More

Beckman Coulter™ SPRIselect™

Product Code. 15676104
Encompass
Change view
Click to view available options
Quantity:
5mL
60mL
Unit Size:
5mL
60mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15676104 5mL 5mL
15605838 60mL 60mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15676104 Supplier Beckman Coulter™ Supplier No. B23317

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purify DNA fragments of the desired size for library preparation. Beckman Coulter™ SPRIselect™ chemistry speeds and simplifies nucleic acid size selection using paramagnetic bead-based chemistry for next-generation sequencing.

Proprietary SPRI paramagnetic bead-based chemistry provides:

  • Tunable fragment size selection to suit specific applications
  • Predictable, consistent size selection between runs and reagent lots
  • Compatibility with manual and automated processing
  • Customizable size selection: from 150 to 800 base pairs
  • Suitable for manual or automated workflows; can be run on Biomek platforms

Specifications

Content And Storage 15-30°C
Form Liquid
Format Liquid
For Use With (Equipment) Automated processing systems, e.g. Biomek
Matrix and Size Standards Tunable size selection from 150 to 800 base pairs
pH 8.0 - 8.4
Quantity 5mL
For Use With (Application) Standard PCR
Sample Type Nucleic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.