missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPRED2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24091-1-AP-150UL
This item is not returnable.
View return policy
Description
SPRED2 is a member of the Sprouty /SPRED family of proteins that regulate growth factor-induced activation of the MAP kinase cascade (Nonami et al., 2004 ).Specifications
| SPRED2 | |
| Polyclonal | |
| Unconjugated | |
| SPRED2 | |
| Spred 2, SPRED2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114716, 200734, 305539 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q3C2P8, Q7Z698, Q924S7 | |
| SPRED2 | |
| SPRED2 Fusion Protein Ag21359 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction