missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SPRED2 Polyclonal Antibody

Product Code. 12808502
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12808502 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12808502 Supplier Invitrogen™ Supplier No. PA520625

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is human small intestine tissue lysate. PA5-20625 can be used with blocking peptide PEP-0745.

SPRED2 is a member of the Sprouty /SPRED family of proteins that regulate growth factor-induced activation of the MAP kinase cascade (Nonami et al., 2004 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPRED2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Spred2
Gene Accession No. Q7Z698
Gene Alias C79158; Spred2; Spred-2; sprouty protein with EVH-1 domain 2, related sequence; sprouty related EVH1 domain containing 2; sprouty related, EVH1 domain containing 2; sprouty-related, EVH1 domain containing 2; sprouty-related, EVH1 domain-containing protein 2
Gene Symbols Spred2
Host Species Rabbit
Immunogen A 13 amino acid peptide near the center of the human Spred2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200734
Target Species Human
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.