missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SPRED2 Polyclonal antibody specifically detects SPRED2 in Human samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | SPRED2 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | DyLight 488 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | FLJ31917, MGC163164, Spred-2related sequence, sprouty-related, EVH1 domain containing 2, sprouty-related, EVH1 domain-containing protein 2 |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a 13 amino acid synthetic peptide near the center of the human Spred2. The immunogen is located within amino acids 140 - 190 of Spred2. Amino Acid Squence: SNSSQKREQPTRT |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?