missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPRED2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76342
This item is not returnable.
View return policy
Description
SPRED2 Polyclonal specifically detects SPRED2 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| SPRED2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| FLJ31917, MGC163164, Spred-2related sequence, sprouty-related, EVH1 domain containing 2, sprouty-related, EVH1 domain-containing protein 2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This Spred2 antibody is predicted to have no cross-reactivity to Spred1 or Spred3. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000 | |
| NP_861449 | |
| SPRED2 | |
| Antibody was raised against a 13 amino acid synthetic peptide near the center of the human Spred2. The immunogen is located within amino acids 140 - 190 of Spred2. Amino Acid Squence: SNSSQKREQPTRT | |
| 0.1 mg | |
| Signal Transduction | |
| 200734 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction