missing translation for 'onlineSavingsMsg'
Learn More

SPRED2 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18426696 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18426696 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18426696 Supplier Novus Biologicals Supplier No. NBP176342AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SPRED2 Polyclonal antibody specifically detects SPRED2 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPRED2
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias FLJ31917, MGC163164, Spred-2related sequence, sprouty-related, EVH1 domain containing 2, sprouty-related, EVH1 domain-containing protein 2
Host Species Rabbit
Immunogen Antibody was raised against a 13 amino acid synthetic peptide near the center of the human Spred2. The immunogen is located within amino acids 140 - 190 of Spred2. Amino Acid Squence: SNSSQKREQPTRT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 200734
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.