missing translation for 'onlineSavingsMsg'
Learn More

SPRED1 Antibody (M23-P2G3), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-50308V

Product Code. 18172489

  • £360.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SPRED1 Monoclonal specifically detects SPRED1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SPRED1
Monoclonal
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
SPRED1
Peptide Sequence YVPLRMCHR (amino acids 420 - 428)
0.1 mL
Primary
Human
Purified
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
M23-P2G3
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
EVH1/Sprouty domain containing protein, FLJ33903, hSpred1, NFLS, spred-1, sprouty-related, EVH1 domain containing 1, sprouty-related, EVH1 domain-containing protein 1, suppressor of Ras/MAPK activation
Mouse
Protein A purified
RUO
161742
Store at 4C in the dark.
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.