missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPRED1 Antibody (M23-P2G3), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50308MFV500
This item is not returnable.
View return policy
Description
SPRED1 Monoclonal antibody specifically detects SPRED1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| SPRED1 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry | |
| M23-P2G3 | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
| EVH1/Sprouty domain containing protein, FLJ33903, hSpred1, NFLS, spred-1, sprouty-related, EVH1 domain containing 1, sprouty-related, EVH1 domain-containing protein 1, suppressor of Ras/MAPK activation | |
| Peptide Sequence YVPLRMCHR (amino acids 420 - 428) | |
| 0.1 mL | |
| Signal Transduction | |
| 161742 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction