Learn More
GVS SPR492 Pair of ABEK1P3 Replacement Filters
Description
These filters are recommended for multigas and dust risks, in chemical production and handling environment. Suitable all types of organic gases, inorganic gases, formaldehyde, chemical gases, dust and ammonia.
- Extremely lightweight and compact to give the wearer as much comfort as possible.
- It offers a full range of vision without interfering with other eye or ear protection which users are required to wear.
- Low breathing resistance to reduce the risk of user fatigue and to add additional comfort.
- Filters last up to 3 years and can be easily replaced.
- Made from soft thermoplastic odorless elastomer that is hypoallergenic (without latex and silicone) and an easy-to-adjust headband.
- Certifications: Mask Body EN140: 1998. ABEK1P 3 R D Filters EN14387: 2004 + A1: 2008. CE 2797
Specifications
Specifications
| Color | White |
| Filter Type | ABEK1P3 |
| Product Type | Filter |
| With Exhalation Valve | No |