missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SPR Recombinant Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
500 μg
Unit Size:
100µg
500µg
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-262 of Mouse SPR The Recombinant Mouse SPR Protein is derived from E. coli. The Recombinant Mouse SPR Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Concentration | 1mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | 20mM Tris-HCl buffer (pH8.5) containing 1mM DTT, 10% glycerol containingno Preservative |
| Gene ID (Entrez) | 6697 |
| Name | Recombinant SPR Protein |
| Purification Method | >95%, by SDS-PAGE |
| Quantity | 500 μg |
| Source | E. coli |
| Storage Requirements | Store at 4°Cshort term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. |
| Regulatory Status | RUO |
| Show More |
For Research Use Only (RUO)
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction