missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ SPR Recombinant Protein

Product Code. 18215681 Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
500 μg
Unit Size:
100µg
500µg
This item is not returnable. View return policy

Product Code. 18215681

Brand: Novus Biologicals™ NBP260206100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-262 of Mouse SPR The Recombinant Mouse SPR Protein is derived from E. coli. The Recombinant Mouse SPR Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1mg/mL
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.5) containing 1mM DTT, 10% glycerol containingno Preservative
Gene ID (Entrez) 6697
Name Recombinant SPR Protein
Purification Method >95%, by SDS-PAGE
Quantity 100 μg
Source E. coli
Storage Requirements Store at 4°Cshort term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Alias EC 1.1.1.153, SDR38C1, sepiapterin reductase, sepiapterin reductase (7,8-dihydrobiopterin:NADP+ oxidoreductase), short chain dehydrogenase/reductase family 38C, member 1
Gene Symbol Spr
Product Type Recombinant Protein
Recombinant Recombinant
Show More Show Less

For Research Use Only (RUO)

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.