missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPR Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA526949
This item is not returnable.
View return policy
Description
SPR Polyclonal Antibody for Western Blot
SPR encodes an aldo-keto reductase that catalyzes the NADPH-dependent reduction of pteridine derivatives and is important in the biosynthesis of tetrahydrobiopterin (BH4). Mutations in this gene result in DOPA-responsive dystonia due to sepiaterin reductase deficiency. A pseudogene has been identified on chromosome 1.
Specifications
| SPR | |
| Polyclonal | |
| Unconjugated | |
| SPR | |
| AA409688; ENSMUSG00000071345; Gm10328; SDR38C1; sepiapterin reductase; sepiapterin reductase (7,8-dihydrobiopterin:NADP+ oxidoreductase); short chain dehydrogenase/reductase family 38C, member 1; SPR | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 20751, 6697 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.25 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P35270, Q64105 | |
| SPR | |
| KLH conjugated synthetic peptide between 231-260 amino acids from the C-terminal region of human SPR. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction