missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPPL3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113253
This item is not returnable.
View return policy
Description
SPPL3 Polyclonal Antibody for ELISA, Western Blot
Signal peptide peptidase (SPP) is an aspartyl protease that mediates clearance of signal peptides by proteolysis within the endoplasmic reticulum (ER). Like presenilins, SPP contains a critical GXGD motif in its C-terminal catalytic center. SPPL3 is one of several presenilin homologues/SPP-like proteins (PSHs/SPPL) that have been identified.
Specifications
| SPPL3 | |
| Polyclonal | |
| Unconjugated | |
| SPPL3 | |
| 4833416I09Rik; fb94d08; Imp2; IMP-2; Intramembrane protease 2; MDHV1887; Presenilin homologous protein 1; presenilin-like protein 4; PRO4332; PSH1; PSL4; putative intramembrane aspartyl protease; RGD1562826; signal peptide peptidase 3; signal peptide peptidase like 3; signal peptide peptidase-like 3; signal peptide peptidase-like 3, pseudogene 1; Signal peptide peptidase-like 3-like protein; signal peptide peptidase-like protein 3; Sppl3; sppl3 protein; Sppl3-ps1; SPP-like 3; SPP-like 3 protein; upregulated during skeletal muscle growth 3; Usmg3; wu:fb94d08; zgc:114093 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 121665, 360822, 74585 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 0.99 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8TCT6, Q9CUS9 | |
| SPPL3 | |
| Recombinant Human Signal peptide peptidase-like 3 protein (213-263AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction