missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPPL2a Antibody [Janelia Fluor™ 669], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-45830JF669
This item is not returnable.
View return policy
Description
SPPL2a Polyclonal antibody specifically detects SPPL2a in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| SPPL2a | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot | |
| Janelia Fluor 669 | |
| EC 3.4.23, EC 3.4.23.-, IMP-3, IMP3intramembrane cleaving protease, Intramembrane protease 3, Presenilin-like protein 2, PSL2, signal peptide peptidase-like 2A, SPPL2a, SPP-like 2A | |
| signal peptide peptidase-like 2A. (Uniprot# Q8TCT8) | |
| 0.1 mL | |
| Cell Biology | |
| 84888 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction