missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD336 (NKp44) Mouse anti-Human, BUV615, Clone: p44-8, BD OptiBuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 752353
This item is not returnable.
View return policy
Description
CD336 (NKp44) Monoclonal antibody specifically detects CD336 (NKp44) in Human samples. It is validated for Flow Cytometry.
Specifications
| CD336 (NKp44) | |
| Monoclonal | |
| 0.2 mg/mL | |
| Hu NKp44 (CD336) BUV615 P44-8 50 μg | |
| Human NKp44 | |
| RUO | |
| CD336 (NKp44) | |
| Refrigerate (2 to 8°C) | |
| Human |
| Flow Cytometry | |
| p44-8 | |
| Brilliant Ultraviolet 615 | |
| Mouse | |
| 1 x 50 μg | |
| Primary | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction