missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPOUT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15653
This item is not returnable.
View return policy
Description
SPOUT1 Polyclonal specifically detects SPOUT1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| C9orf114 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| CENP-32, centromere protein 32, chromosome 9 open reading frame 114, DKFZp566D143, HSPC109, MGC29492 | |
| Rabbit | |
| 42 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| Q5T280 | |
| C9orf114 | |
| Recombinant protein encompassing a sequence within the center region of human C9orf114. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 51490 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction