missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPOUT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35850-100ul
This item is not returnable.
View return policy
Description
SPOUT1 Polyclonal antibody specifically detects SPOUT1 in Human,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| SPOUT1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| CENP-32, centromere protein 32, chromosome 9 open reading frame 114, DKFZp566D143, HSPC109, MGC29492 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 163-340 of human SPOUT1 (NP_057474.2).,, Sequence:, HQDLQFAGLLNPLDSPHHMRQDEESEFREGIVVDRPTRPGHGSFVNCGMKKEVKIDKNLEPGLRVTVRLNQQQHPDCKTYHGKVVSSQDPRTKAGLYWGYTVRLASCLSAVFAEAPFQDGYDLTIGTSERGSDVASAQLPNFRHALVVFGGLQGLEAGADADPNLEVAEPSVLFDLYV | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 51490 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction