missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPOP Rabbit anti-Human, Clone: 2N8L8, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | SPOP |
|---|---|
| Clone | 2N8L8 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18340656
|
Novus Biologicals
NBP3-15784-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18330587
|
Novus Biologicals
NBP3-15784-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SPOP Monoclonal antibody specifically detects SPOP in Human samples. It is validated for Western BlotSpecifications
| SPOP | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| HIB homolog 1, Roadkill homolog 1, speckle-type POZ protein, TEF2 | |
| A synthetic peptide corresponding to a sequence within amino acids 250-350 of human SPOP (O43791). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 2N8L8 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 8405 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title