Learn More
Invitrogen™ SPOP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113929
Description
Immunogen sequence: YHASDVLETS GWKSMVVSHP HLVAEAYRSL ASAQCPFLGP PRKRLKQS For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
Component of a cullin-RING-based BCR (BTB-CUL3-RBX1) E3 ubiquitin-protein ligase complex that mediates the ubiquitination of target proteins, leading most often to their proteasomal degradation.
Specifications
| SPOP | |
| Polyclonal | |
| Unconjugated | |
| SPOP | |
| AI315626; BTBD32; HIB homolog 1; Pcif1; PDX-1 C-terminal-interacting factor 1; roadkill homolog 1; similar to speckle-type POZ protein; speckle type BTB/POZ protein; speckle type POZ protein; speckle-type POZ protein; Spop; SPOP1; TEF2; zgc:64140 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 8405 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O43791 | |
| SPOP | |
| Synthetic peptide directed towards the C terminal region of human SPOP. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.