missing translation for 'onlineSavingsMsg'
Learn More

Sponge Nickel, A-5000, nonpromoted, Thermo Scientific Chemicals

Product Code. 11396279
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11396279 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11396279 Supplier Thermo Scientific Alfa Aesar Supplier No. 044867.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Slurry in water

Hydrogenation of nitro groups to amines, hydrogenation of carbonyl groups, hydrogenation of olefins to alkanes, reductive alkylation (stereoselective)Sponge Nickel, A-5000, nonpromoted is a standard catalyst used for most hydrogenation reactions such as hydrogenation of xylose to xylitol. It is also involved in the industrial hydrogenation of butyraldehyde to butanol, 2,4-dinitrotoluene to 2,4-diaminotoluene. Further, it is used as a catalyst in reductive alkylation reaction, especially tertiary diemethylaliphatic amine from aliphatic primary amine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Color Silver
Appearance Silver colored sponge
Quantity 500 g
Odor Odorless
Sensitivity Air Sensitive
Form Slurry
Solubility Information Insoluble in water.
Chemical Name or Material Nickel, Nonpromoted

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.