missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPON2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20513-1-AP-20UL
This item is not returnable.
View return policy
Description
Cell adhesion protein that promotes adhesion and outgrowth of hippocampal embryonic neurons. Binds directly to bacteria and their components and functions as an opsonin for macrophage phagocytosis of bacteria. Essential in the initiation of the innate immune response and represents a unique pattern-recognition molecule in the ECM for microbial pathogens. Binds bacterial lipopolysaccharide (LPS).Specifications
| SPON2 | |
| Polyclonal | |
| Unconjugated | |
| SPON2 | |
| DIL 1, DIL1, M spondin, Mindin, SPON2, Spondin 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10417 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.41 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9BUD6 | |
| SPON2 | |
| SPON2 Fusion Protein Ag14337 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction