missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPON2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA537919
This item is not returnable.
View return policy
Description
This antibody is predicted to react with mouse, rat and dog based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
Cell adhesion protein that promotes adhesion and outgrowth of hippocampal embryonic neurons. Binds directly to bacteria and their components and functions as an opsonin for macrophage phagocytosis of bacteria. Essential in the initiation of the innate immune response and represents a unique pattern-recognition molecule in the ECM for microbial pathogens. Binds bacterial lipopolysaccharide (LPS).
Specifications
| SPON2 | |
| Polyclonal | |
| Unconjugated | |
| SPON2 | |
| 2310045I24Rik; AI504350; differentially expressed in cancerous and non-cancerous lung cells 1; DIL1; DIL-1; DKFZp686G21139; FLJ16313; FLJ22401; FLJ34460; F-spondin; MINDIN; Mspondin; M-SPONDIN; SPON2; spondin 2; spondin 2, extracellular matrix protein; spondin-2; UNQ435/PRO866 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 100689, 10417, 171569 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q8BMS2, Q9BUD6, Q9WV75 | |
| SPON2 | |
| Peptide with sequence C-RTRYVRVQPANNGSP, from the internal region near the C-terminus of SPON2 (aa301-315). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction