missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPON2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566822
This item is not returnable.
View return policy
Description
Immunogen sequence: HSLVSFVVRIV PSPDWFVGVD SLDLCDGDRW REQAALDLYP YDAGTDSGFT FSSPNFATIP QDTVTEITSS SP Highest antigen sequence identity to the following orthologs - mouse 89%, rat 89%.
Cell adhesion protein that promotes adhesion and outgrowth of hippocampal embryonic neurons. Binds directly to bacteria and their components and functions as an opsonin for macrophage phagocytosis of bacteria. Essential in the initiation of the innate immune response and represents a unique pattern-recognition molecule in the ECM for microbial pathogens. Binds bacterial lipopolysaccharide (LPS).
Specifications
| SPON2 | |
| Polyclonal | |
| Unconjugated | |
| SPON2 | |
| 2310045I24Rik; AI504350; differentially expressed in cancerous and non-cancerous lung cells 1; DIL1; DIL-1; DKFZp686G21139; FLJ16313; FLJ22401; FLJ34460; F-spondin; MINDIN; Mspondin; M-SPONDIN; SPON2; spondin 2; spondin 2, extracellular matrix protein; spondin-2; UNQ435/PRO866 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10417 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BUD6 | |
| SPON2 | |
| Recombinant Human SPON2. Recombinant protein control fragment (Product #RP-105379). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction