missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPO11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-58172
This item is not returnable.
View return policy
Description
SPO11 Polyclonal specifically detects SPO11 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| SPO11 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| Cancer/testis antigen 35, CT35MGC39953, meiotic recombination protein SPO11, SPO11 meiotic protein covalently bound to DSB homolog (S. cerevisiae), SPO11 meiotic protein covalently bound to DSB-like (S. cerevisiae), SPO11, meiotic protein covalently bound to DSB (S. cerevisiae)-like | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Bovine: 100%; Canine: 100%; Guinea pig: 100%; Human: 100%; Pig: 100%; Equine: 92%; Mouse: 92%; Rabbit: 92%; Rat: 92%; Chicken: 84%; Zebrafish: 76%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9Y5K1 | |
| SPO11 | |
| Synthetic peptides corresponding to SPO11(SPO11 meiotic protein covalently bound to DSB homolog (S. cerevisiae)) The peptide sequence was selected from the N terminal of SPO11. Peptide sequence KFSLILKILSMIYKLVQSNTYATKRDIYYTDSQLFGNQTVVDNIINDISC The peptide sequence for this immunogen was taken from within the described region. | |
| 100 μL | |
| DNA Repair, Editing and Processing Endonucleases | |
| 23626 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction