missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPNS1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£150.00 - £417.00
Specifications
| Antigen | SPNS1 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18650448
|
Novus Biologicals
NBP2-41252-0.025mg |
0.025 mg |
£150.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18115319
|
Novus Biologicals
NBP2-41252 |
0.1 mg |
£417.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SPNS1 Polyclonal specifically detects SPNS1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| SPNS1 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| 83985 | |
| Antibody was raised against an 18 amino acid peptide near the amino terminus of human SPINSTER. The immunogen is located within the first 50 amino acids of SPINSTER. Amino Acid Squence: STGNPKSEEPEVPDQEGL | |
| Primary | |
| SPINSTER antibody is human specific. At least four isoforms of SPINSTER are known to exist. This antibody is predicted to not cross-react with other members of the spinster family of proteins. |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Human | |
| FLJ38358, HSpin1Spinster-like protein 1, LAT, PP2030, protein spinster homolog 1, SPIN1nrs, SPINL, spinster homolog 1 (Drosophila) | |
| SPNS1 | |
| IgG | |
| Affinity Purified | |
| 56 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title