missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Splunc2 Monoclonal Antibody (3F8E4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531653
This item is not returnable.
View return policy
Description
MA5-31653 has been tested in indirect ELISA.
SPLUNC2 is one member of PLUNC family, the gene undergoes alternative splicing using two 5' non-coding exons, suggesting that the gene is regulated by alternative promoters.multiple SPLUNC2 isoforms are found in the oral cavity and suggest that these proteins may be differentially regulated in distinct tissues where they may function in the innate immune response. Mucin plugs, mucous and intermediate cells of mucoepidermoid carcinomas were positive for LPLUNC1 and SPLUNC2, but areas composed of epidermoid and clear cells were negative for all PLUNCs. Papillary cystadenocarcinoma was positive for all PLUNCs.
Specifications
| Splunc2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P07743, Q96DR5 | |
| BPIFA2 | |
| Purified recombinant fragment of mouse mSplunc2 (AA: 16-169) expressed in E. coli. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 3F8E4 | |
| Unconjugated | |
| BPIFA2 | |
| bA49G10.1; BPI fold containing family A member 2; BPI fold containing family A, member 2; BPI fold containing family A, member 2E; BPI fold-containing family A member 2; BPIFA2; Bpifa2e; C20orf70; mSplunc2; parotid secretory protein; PSP; short palate; short palate, lung and nasal epithelium carcinoma-associated protein 2; SPLUNC2; UNQ510/PRO1025 | |
| Mouse | |
| Protein G | |
| RUO | |
| 140683, 19194 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction