missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Split/Splitless Liners for Agilent™ 6890 GCs: Low Pressure Drop

Product Code. 13194412
Encompass_Preferred
Change view
Click to view available options
Product Type:
IP Low Pressure Drop Inlet Liner
IP Precision Low Pressure Drop Inlet Liner
Deactivation:
IP Deactivation
Siltek Deactivation
Quantity:
1 Ea.
5/Pk.
Unit Size:
Pack of 5
Product Code. Product Type Deactivation Quantity unitSize
13194412 IP Precision Low Pressure Drop Inlet Liner Siltek Deactivation 5/Pk. Pack of 5
13114412 IP Precision Low Pressure Drop Inlet Liner IP Deactivation 1 Ea. -
13164412 IP Precision Low Pressure Drop Inlet Liner IP Deactivation 5/Pk. -
10530322 IP Precision Low Pressure Drop Inlet Liner IP Deactivation 5/Pk. -
10025473 IP Low Pressure Drop Inlet Liner IP Deactivation 5/Pk. Pack of 5
13194332 IP Low Pressure Drop Inlet Liner IP Deactivation 5/Pk. Pack of 5
6 options
This item is not returnable. View return policy

Product Code. 13194412

Brand: Restek™ 21033213.5

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes wool packing

  • I.D. 4mm

Specifications

Deactivation Siltek Deactivation
Quantity 5/Pk.
Length (Metric) 7.85 cm
Packing Material Deactivated Wool
Product Type IP Precision Low Pressure Drop Inlet Liner
Diameter (Metric) Inner 0.4 cm
Diameter (Metric) Outer 0.63 cm
For Use With (Equipment) Agilent GCs Equipped with Split/Splitless Inlets
Liner Material Borosilicate Glass
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.