missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Split/Splitless Liners for Agilent™ 6890 GCs: Low Pressure Drop

Includes wool packing

Brand:  Restek™ 21032

Code : Z3

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 13114412

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • I.D. 4mm

Specifications

Specifications

Deactivated Wool
1 Ea.
0.4 cm
Agilent GCs Equipped with Split/Splitless Inlets
IP Precision Low Pressure Drop Inlet Liner
7.85 cm
0.63 cm
Borosilicate Glass
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers