missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical™ PerkinElmer Split / Splitless FocusLiners
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
£147.00 - £700.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12914031
|
Trajan Scientific and Medical™
09209225 |
25/Pk. |
£700.00
Pack of 5 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||||
10309142
|
Trajan Scientific and Medical™
092059 |
5/Pk. |
£147.00
Pack of 5 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||||
Description
- Each batch of liners tested for activity
- Manufactured from borosilicate glass
- Color coded by geometry for easy identification
- For use with PerkinElmer instruments