Learn More
Lenz Laborglasintrumente Splash Heads Acc. to Stutzer, Bent
Product Code. p-7222613
Click to view available options
Standard Taper Joint:
Cone Vertical NS 14/23, Cone Bent NS 14/23
Cone Vertical NS 24/29, Cone Bent NS 19/26
Cone Vertical NS 24/29, Cone Bent NS 24/29
Unit Size:
Each
Product Code. 10717711
Brand: Lenz Laborglasintrumente 05037011
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Bent shape (75°), with two NS cones
  • For separation of drops (direct placement onto distillation flasks)
  • Made of DURAN™ tubing
Specifications
Material Duran Glass
Standard Taper Joint Cone Vertical NS 14/23, Cone Bent NS 14/23