missing translation for 'onlineSavingsMsg'
Learn More

SPK-601, MedChemExpress

Product Code. p-200101861
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16434097 50 mg 50mg
16428198 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16434097 Supplier Medchem Express Supplier No. HY70083

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SPK-601 (LMV-601) is an inhibitor of the phosphatidylcholine-specific phospholipase C (PC-PLC). SPK-601 also can be used as an antimicrobial agent.

SPK-601 (LMV-601) is an inhibitor of the phosphatidylcholine-specific phospholipase C (PC-PLC). SPK-601 also can be used as an antimicrobial agent.

Specifications

Chemical Name or Material SPK-601
Physical Form Solid
CAS 1096687-52-3
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C11H15KOS2
For Use With (Application) COVID-19-anti-virus
Synonym LMV-601
Solubility Information DMSO : 5 mg/mL (18.76 mM; Need ultrasonic)
SMILES [H][C@]12[C@@]3([C@H](C[C@]([H])([C@@]1([H])CCC2)C3)OC(S[K])=S)[H]
Molecular Weight (g/mol) 266.46
Formula Weight 266.46
Percent Purity 98.06%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.