missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPK-601, MedChemExpress
Description
SPK-601 (LMV-601) is an inhibitor of the phosphatidylcholine-specific phospholipase C (PC-PLC). SPK-601 also can be used as an antimicrobial agent.
Specifications
Specifications
| Chemical Name or Material | SPK-601 |
| Physical Form | Solid |
| CAS | 1096687-52-3 |
| Color | Off-White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C11H15KOS2 |
| For Use With (Application) | COVID-19-anti-virus |
| Synonym | LMV-601 |
| Solubility Information | DMSO : 5 mg/mL (18.76 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?