missing translation for 'onlineSavingsMsg'
Learn More

Spirotetramat Metabolite BYI08330-mono-hydroxy, TRC

Product Code. 30135061
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135061 50 mg 50mg
17943735 5 mg 5mg
17951795 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135061 Supplier Toronto Research Chemicals Supplier No. TRCS68331050MG
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1172134-12-1
Molecular Formula C18 H25 N O3
Molecular Weight (g/mol) 303.4
Synonym stereoisomer of 3-(2,5-Dimethylphenyl)-4-hydroxy-8-methoxy-1-azaspiro[4.5]decan-2-one, Spirotetramat-mono-hydroxy
IUPAC Name 3-(2,5-dimethylphenyl)-4-hydroxy-8-methoxy-1-azaspiro[4.5]decan-2-one
SMILES CO[C@@H]1CC[C@@]2(CC1)NC(=O)C(C2O)c3cc(C)ccc3C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C18H25NO3/c1-11-4-5-12(2)14(10-11)15-16(20)18(19-17(15)21)8-6-13(22-3)7-9-18/h4-5,10,13,15-16,20H,6-9H2,1-3H3,(H,19,21)/t13-,15?,16?,18+
Formula Weight 303.1834
Percent Purity >95%
Chemical Name or Material Spirotetramat-mono-hydroxy
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.