missing translation for 'onlineSavingsMsg'
Learn More

Spironolactone, TRC

Product Code. 30420405
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30420405 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30420405 Supplier Toronto Research Chemicals Supplier No. TRCS68300050MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Spironolactone
CAS 52-01-7
Molecular Formula C24 H32 O4 S
Quantity 50 mg
Synonym Spironolactone, (2'R)-7α-(Acetylsulfanyl)-3',4'-dihydro-5'H-spiro[androst-4-ene-17,2'-furan]-3,5'-dione, Pregn-4-ene-21-carboxylic acid','7-(acetylthio)-17-hydroxy-3-oxo-','γ-lactone','(7α,17α)-, 17α-Pregn-4-ene-21-carboxylic acid','17-hydroxy-7α-mercapto-3-oxo-','γ-lactone','acetate (6CI,8CI), Spiro[17H-cyclopenta[a]phenanthrene-17,2'(5'H)-furan]','pregn-4-ene-21-carboxylic acid deriv., 17-Hydroxy-7α-mercapto-3-oxo-17α-pregn-4-ene-21-carboxylic acid γ-lactone 7-acetate, 3-(3-Oxo-7α-acetylthio-17β-hydroxy-4-androsten-17α-yl)propionic acid-γ-lactone, 3-(3-keto-7α-Acetylthio-17β-hydroxy-4-androsten-17α-yl)propionic acid lactone, 3'-(3-Oxo-7α-acetylthio-17β-hydroxyandrost-4-en-17α-yl)-propionic acid lactone, 7α-(Acetylthio)-17-hydroxy-3-oxo-17α-pregn-4-ene-21-carboxylic acid γ-lactone, 7α-Acetylthio-3-oxo-17α-pregn-4-ene-21,17β-carbolactone, Abbolactone, Aldace, Aldactone, Aldactone A, Aldopur, Almatol, Altex, Aquareduct, Berlactone, Deverol, Diatensec, Dira, Duraspiron, Euteberol, Lacalmin, Lacdene, Laractone, NSC 150399, Nefurofan, Novo-Spiroton, Osiren, Osyrol, Quimolactona, SC 9420, Sagisal, Sincomen, Spiresis, Spiretic, Spiridon, Spiro-Tablinen, Spiroctan, Spiroderm, Spirolactone, Spirolang, Spirolone, Spirone, Spironol, Spironolactone A, Supra-Puren, Suracton, Uractone, Urusonin, Verospiron, Verospirone, Xenalon
InChI Formula InChI=1S/C24H32O4S/c1-14(25)29-19-13-15-12-16(26)4-8-22(15,2)17-5-9-23(3)18(21(17)19)6-10-24(23)11-7-20(27)28-24/h12,17-19,21H,4-11,13H2,1-3H3/t17-,18-,19+,21+,22-,23-,24+/m0/s1
SMILES CC(=O)S[C@@H]1CC2=CC(=O)CC[C@]2(C)[C@H]3CC[C@@]4(C)[C@@H](CC[C@@]45CCC(=O)O5)[C@H]13
IUPAC Name S-[(7R,8R,9S,10R,13S,14S,17R)-10,13-dimethyl-3,5'-dioxospiro[2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene-17,2'-oxolane]-7-yl] ethanethioate
Molecular Weight (g/mol) 416.57
Formula Weight 416.2021
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.