missing translation for 'onlineSavingsMsg'
Learn More

Spironolactone, MedChemExpress

Product Code. p-200099108
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16480998

Brand: Medchem Express HYB0561

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Spironolactone (SC9420) is an orally active aldosterone mineralocorticoid receptor antagonist with an IC50 of 24 nM. Spironolactone is also a potent antagonist of androgen receptor with an IC50 of 77 nM. Spironolactone promotes autophagy in podocytes.

Spironolactone (SC9420) is an orally active aldosterone mineralocorticoid receptor antagonist with an IC50 of 24 nM. Spironolactone is also a potent antagonist of androgen receptor with an IC50 of 77 nM. Spironolactone promotes autophagy in podocytes.

Specifications

Chemical Name or Material Spironolactone
CAS 52-01-7
Color White
Purity Grade Notes Research
Molecular Formula C24H32O4S
Quantity 5 g
Synonym SC9420
Solubility Information DMSO : ≥ 50 mg/mL (120.03 mM) ∣H2O : 0.1 mg/mL (0.24 mM; Need ultrasonic)
SMILES C[C@@]12[C@](OC3=O)(CC3)CC[C@@]1([H])[C@@]([C@@H](CC4=CC5=O)SC(C)=O)([H])[C@]([C@]4(CC5)C)([H])CC2
Molecular Weight (g/mol) 416.57
Formula Weight 416.57
Percent Purity 99.58%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.