missing translation for 'onlineSavingsMsg'
Learn More

Spirodiclofen, MedChemExpress

Product Code. 16488466
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488466 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16488466 Supplier Medchem Express Supplier No. HYB0826

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Spirodiclofen is a broad spectrum acaricide acting via lipid biosynthesis inhibition (LBI) with no cross resistance to currently available acaricides and with additional insecticidal properties.

Spirodiclofen is a broad spectrum acaricide acting via lipid biosynthesis inhibition (LBI) with no cross resistance to currently available acaricides and with additional insecticidal properties.

Specifications

Chemical Name or Material Spirodiclofen
CAS 148477-71-8
Color White
Purity Grade Notes Research
Molecular Formula C21H24Cl2O4
Quantity 100 mg
Synonym BAJ-2740
Solubility Information DMSO : 120 mg/mL (291.74 mM; Need ultrasonic) ∣Ethanol : 120 mg/mL (291.74 mM; Need ultrasonic)
SMILES CCC(C)(C)C(OC1=C(C2=CC=C(Cl)C=C2Cl)C(OC13CCCCC3)=O)=O
Molecular Weight (g/mol) 411.32
Formula Weight 411.32
Percent Purity 99.92%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.