missing translation for 'onlineSavingsMsg'
Learn More

Spiro[1,3-dioxolan-2,3′-estra-5′(10′),9′(11′)-dien]-17′-one, Mikromol™

Product Code. 17959679
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17959679 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17959679 Supplier Mikromol Supplier No. MM1274.020025
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Spiro[1,3-dioxolan-2,3′-estra-5′(10′),9′(11′)-dien]-17′-one
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Estra-5(10),9(11)-diene-3,17-dione 3-Ethylene Ketal
CAS 5571-36-8
Analyte or Component Names Estra-5(10),9(11)-diene-3,17-dione 3-Ethylene Ketal
Shipping Condition Room Temperature
Molecular Formula C20 H26 O3
Certifications/Compliance ISO 9001
Synonym Estra-5(10),9(11)-diene-3,17-dione, cyclic 3-(1,2-ethanediyl acetal), Estra-5(10),9(11)-diene-3,17-dione, cyclic 3-(ethylene acetal) (7CI,8CI), Spiro[3H-cyclopenta[a]phenanthrene-3,2'-[1,3]dioxolane], estra-5(10),9(11)-diene-3,17-dione deriv., 3,3-(Ethylenedioxy)estra-5(10),9(11)-dien-17-one, Spiro[1,3-dioxolan-2,3′-estra-5′(10′),9′(11′)-dien]-17′-one, Dienogest Imp. L (EP), Dienogest Impurity L
InChI Formula InChI=1S/C20H26O3/c1-19-8-6-15-14-7-9-20(22-10-11-23-20)12-13(14)2-3-16(15)17(19)4-5-18(19)21/h6,16-17H,2-5,7-12H2,1H3/t16-,17+,19+/m1/s1
SMILES C[C@]12CC=C3[C@@H](CCC4=C3CCC5(C4)OCCO5)[C@@H]1CCC2=O
IUPAC Name (8S,13S,14S)-13-methylspiro[1,2,4,6,7,8,12,14,15,16-decahydrocyclopenta[a]phenanthrene-3,2'-1,3-dioxolane]-17-one
Molecular Weight (g/mol) 314.42
Quantity 25 mg
Formula Weight 314.1882 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.