missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPIRE2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17757-1-AP-150UL
This item is not returnable.
View return policy
Description
Acts as a actin nucleation factor, remains associated with the slow-growing pointed end of the new filament. Involved in vesicle transport processes providing a novel link between actin organization and intracellular transport.Specifications
| SPIRE2 | |
| Polyclonal | |
| Unconjugated | |
| SPIRE2 | |
| KIAA1832, Protein spire homolog 2, Spir 2, SPIR2, spire homolog 2 (Drosophila), SPIRE2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 234857, 307925, 84501 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8K1S6, Q8WWL2 | |
| SPIRE2 | |
| SPIRE2 Fusion Protein Ag12117 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction