missing translation for 'onlineSavingsMsg'
Learn More

SPIRE2 Antibody, Novus Biologicals™

Product Code. p-200072856 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mL
0.25 mg, 25 μL
Unit Size:
0.10mL
25µL
This item is not returnable. View return policy

Product Code. 18109308

Brand: Novus Biologicals NBP247261

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SPIRE2 Polyclonal specifically detects SPIRE2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPIRE2
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:2500 - 1:5000, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:2500 - 1:5000
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias KIAA1832, MGC117166, protein spire homolog 2, Spir-2, SPIR2, spire homolog 2 (Drosophila)
Gene Symbols SPIRE2
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: SLPCILNACSGDAKSTSCINLSVTDAGGSAQRPRPRVLLKAPTLAEMEEMNTSEEEESPCGEVTLKRDRSFSEHDLAQ
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84501
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.