missing translation for 'onlineSavingsMsg'
Learn More

Spiramycin, MedChemExpress

Product Code. 16404396
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16404396 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16404396 Supplier Medchem Express Supplier No. HY100593
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Spiramycin (Rovamycin) is a macrolide antibiotic produced by Streptomyces ambofaciens with against bacteria and Toxoplasma gondii activities, and also has antiparasitic effect. Spiramycin is composed of a 16-member lactone ring, on which three sugars (mycaminose, forosamine, and mycarose) are attached.

Spiramycin (Rovamycin) is a macrolide antibiotic produced by Streptomyces ambofaciens with against bacteria and Toxoplasma gondii activities, and also has antiparasitic effect. Spiramycin is composed of a 16-member lactone ring, on which three sugars (mycaminose, forosamine, and mycarose) are attached.

Specifications

Chemical Name or Material Spiramycin
Physical Form Powder
CAS 8025-81-8
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C43H74N2O14
For Use With (Application) COVID-19-immunoregulation
Synonym Rovamycin
Solubility Information DMSO : ≥ 100 mg/mL (118.62 mM)
SMILES O[C@H](C1)[C@H](OC)[C@@H](O[C@H]2[C@H](O)[C@@H](N(C)C)[C@H](O[C@@H]3O[C@@H](C)[C@H](O)[C@@](O)(C)C3)[C@@H](C)O2)[C@@H](CC=O)C[C@@H](C)[C@@H](O[C@@H]4O[C@H](C)[C@@H](N(C)C)CC4)/C=C/C=C/C[C@@H](C)OC1=O
Molecular Weight (g/mol) 843.05
Formula Weight 843.05
Percent Purity 97.87%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.