missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Spiral Screw for PowderProof™ Sampler

Buerkle™ Spiral Screw for PowderProof™ Sampler is a spare part for use with Buerkle™ PowderProof™ Sampler.

Brand:  Buerkle™ 5302-0312

Code : A0

Additional Details : Weight : 0.23000kg

 View more versions of this product

Product Code. 11817383

  • £404.00 / Each
Estimated Shipment: 13-06-2024
to see stock.
Add to basket

Description

Description

  • Spare part for Buerkle™ PowderProof™ Sampler
Specifications

Specifications

Spiral Screw
Stainless Steel (V4A)
PowderProof for Powders up to 2 mm dia.
30 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers