Learn More
Buerkle™ Spiral Screw for PowderProof™ Sampler
Product Code. 11887383 Shop All Bürkle Products
Change view
Click to view available options
Length (Metric):
90 cm
Unit Size:
Each
Product Code. Length (Metric) unitSize
11887383 90 cm Each
1 options
Product Code. 11887383 Supplier Buerkle™ Supplier No. 53020912
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
PowderProof is the practically proven professional solution for mobile sampling of powders and granules. Different lances and augers can be attached to the robust drive unit and swapped without difficulty. Stainless steel V4A (1.4404)
PowderProof is the practically proven professional solution for mobile sampling of powders and granules. Different lances and augers can be attached to the robust drive unit and swapped without difficulty. Stainless steel V4A (1.4404)
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel (V4A)
For Use With (Equipment) For up to 2 mm dia. Powders
Length (Metric) 90 cm